Learn More
Beckman Coulter Tube Holder Adapter for 14 mm Diameter Tubes
Catalog No. NC3920959
Encompass_Preferred
The Beckman Coulter D30246 tube adapter enables 14 mm diameter tubes to be used in compatible Beckman tube holders and instruments, providing a secure and centered fit so smaller tubes can be processed in equipment designed for larger tube sizes.
  • Accommodates tubes with a 14 mm outer diameter.
  • Provides a secure fit in compatible tube holders.
  • Enables use of smaller tubes in instruments designed for larger diameters.
  • Simplifies sample handling and setup for routine laboratory workflows.
  • Reusable design suitable for repeated use in standard lab applications.
Catalog No. NC3920959 Supplier Beckman Coulter Supplier No. D30246
The Beckman Coulter D30246 tube adapter enables 14 mm diameter tubes to be used in compatible Beckman tube holders and instruments, providing a secure and centered fit so smaller tubes can be processed in equipment designed for larger tube sizes.
  • Accommodates tubes with a 14 mm outer diameter.
  • Provides a secure fit in compatible tube holders.
  • Enables use of smaller tubes in instruments designed for larger diameters.
  • Simplifies sample handling and setup for routine laboratory workflows.
  • Reusable design suitable for repeated use in standard lab applications.