Learn More
Beckman Coulter Tube Holder Adapter for 38 mm Diameter Tubes
Catalog No. NC3920954
Encompass_Preferred
The Beckman Coulter D17686 tube holder adapter secures laboratory tubes with a 38 mm outer diameter for centrifugation, providing a stable interface for accurate sample processing.
  • Secures tubes with a 38 mm outer diameter.
  • Designed for use in compatible tube carriers or holders.
  • Provides a stable interface for centrifugation applications.
Catalog No. NC3920954 Supplier Beckman Coulter Supplier No. D17686
The Beckman Coulter D17686 tube holder adapter secures laboratory tubes with a 38 mm outer diameter for centrifugation, providing a stable interface for accurate sample processing.
  • Secures tubes with a 38 mm outer diameter.
  • Designed for use in compatible tube carriers or holders.
  • Provides a stable interface for centrifugation applications.