Learn More
Chemglass Life Sciences Tube Holder, Micro, Universal
SDP
Catalog No. 50143320
Encompass
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-143-320 Supplier Chemglass Life Sciences Supplier No. CLS4891U16
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.