Learn More
Fisherbrand™ Tube Holders
Catalog No. 22024239 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
Blood Collection Tube Holder
Blood Collection Tube Holder with Luer Lock Adapter
Blood Transfer Tube Holder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-024-239 Blood Collection Tube Holder
22-024-243 Blood Collection Tube Holder with Luer Lock Adapter
22-024-244 Blood Transfer Tube Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-024-239
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fisherbrand Tube Holders allow for the transfer of blood from a syringe into a blood collection tube.
  • Reduces the risk of transfer-related incident while maintaining specimen viability
Specifications
Disposable Single-use
Autoclavable Not Autoclavable
For Use With (Equipment) Fisherbrand Safety Needles; Standard Multi-Sample Safety Needles
Includes Blood Collection Tube Holder
Latex Free Yes
Sterility Non-sterile