Learn More
Fisherbrand™ Tube Holders
Catalog No. 22024243 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
Blood Collection Tube Holder
Blood Collection Tube Holder with Luer Lock Adapter
Blood Transfer Tube Holder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-024-243 Blood Collection Tube Holder with Luer Lock Adapter
22-024-239 Blood Collection Tube Holder
22-024-244 Blood Transfer Tube Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-024-243
  • PRODUCT PROMOTION
    $83.00
    $27.00 / Pack of 50
    Save $56.00
    67% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • PRODUCT PROMOTION
    $1,315.00
    $432.00 / Case of 16 PK
    Save $883.00
    67% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Fisherbrand Tube Holders allow for the transfer of blood from a syringe into a blood collection tube.
  • Reduces the risk of transfer-related incident while maintaining specimen viability
Specifications
Includes Blood Collection Tube Holder with Luer Lock Adapter
Autoclavable Not Autoclavable
Disposable Single-use
For Use With (Equipment) Fisherbrand Safety Blood Collection Needle
Latex Free Yes
Sterility Sterile