Learn More
Wilmad Labglass TUBE MELTING POINT 1.5X100MM
Catalog No. 50807778
Encompass
Tube, Melting Point, Capillary. Tube O.D.: 1.5mm; Tube O.D.: 1mm; Tube Length: 100mm. Fabricated from low extractable borosilicate glass. Capillary tube is closed at one end and packed in vials containing approximately 100 tubes.
Catalog No. 50-807-778 Supplier Wilmad Labglass Supplier No. LG9060102
Tube, Melting Point, Capillary. Tube O.D.: 1.5mm; Tube O.D.: 1mm; Tube Length: 100mm. Fabricated from low extractable borosilicate glass. Capillary tube is closed at one end and packed in vials containing approximately 100 tubes.