Learn More
Labcon North America SuperClear 1.5mL Screw Cap Microcentrifuge Tubes with Caps, Sterile
Catalog No. NC1587908
Encompass_Preferred
Tube, Microcentrifuge; Screw Cap Style; Labcon; SuperClear; 1.5mL; With attached flat screw caps; Caps include integral ethylene O-ring; Includes molded graduations and labeling spot; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 1.5mL; Natural color caps; 50 tubes per bag/10 bags per pack/10 packs per case; Made in USA
Catalog No. NC1587908 Supplier Labcon North America Supplier No. 36118750009
Tube, Microcentrifuge; Screw Cap Style; Labcon; SuperClear; 1.5mL; With attached flat screw caps; Caps include integral ethylene O-ring; Includes molded graduations and labeling spot; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 1.5mL; Natural color caps; 50 tubes per bag/10 bags per pack/10 packs per case; Made in USA