Learn More
Med Vet International McKesson Conical Bottom Microcentrifuge Tube, 2 mL, 177-111574, 500/Bag
Catalog No. 503044097
Encompass_Preferred
McKesson Microcentrifuge Tube Conical Bottom Plain 2 mL Without Color Coding Snap Cap Polypropylene Tube Large, black, clearly printed graduations in precise increments of 0.1 mL Frosted writing area on each tube for labeling Compatible for use with all standard and high-capacity centrifuge rotors Can be centrifuged at up to 20,000 RCF Quality, food-grade construction certified to be free from DNase, RNase, pyrogen, and DNA Supplied in resealable, tamper-evident, self-standing bags
Catalog No. 50-304-4097 Supplier Med Vet International Supplier No. 1169387BG
McKesson Microcentrifuge Tube Conical Bottom Plain 2 mL Without Color Coding Snap Cap Polypropylene Tube Large, black, clearly printed graduations in precise increments of 0.1 mL Frosted writing area on each tube for labeling Compatible for use with all standard and high-capacity centrifuge rotors Can be centrifuged at up to 20,000 RCF Quality, food-grade construction certified to be free from DNase, RNase, pyrogen, and DNA Supplied in resealable, tamper-evident, self-standing bags