Learn More
Hach Company Tubes, MICRO DIST Cyanide-1, NaOH trapping solution. 21/pk
Catalog No. NC9680119 Shop All Hach Company Products
Encompass
Tubes for Cyanide MICRO DIST® Distillation System. Distill up to 21 samples in 30 minutes. Cyanide method is USEPA Approved for NPDES and NPDWR. 21/pk
Catalog No. NC9680119 Supplier Hach Company Supplier No. A17001
 Restricted Item
Add to Cart
Add to Cart
Tubes for Cyanide MICRO DIST® Distillation System. Distill up to 21 samples in 30 minutes. Cyanide method is USEPA Approved for NPDES and NPDWR. 21/pk