Learn More
Chemglass Life Sciences Tube, NMR, 3mm, Select Series, With Caps, 7" Long, 500 MHz, Concentricity +- 0.0050mm, Camber +- 0.0130mm
SDP
Catalog No. CS35007
Encompass_Preferred
Tube, NMR, 3mm, Select Series, With Caps, 7" Long, 500 MHz, Concentricity 0.0050mm, Camber 0.0130mm. NORELL Select Series NMR Tubes are specifically designed for high-resolution NMR. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.
Catalog No. CS35007 Supplier Chemglass Life Sciences Supplier No. CS35007
Tube, NMR, 3mm, Select Series, With Caps, 7" Long, 500 MHz, Concentricity 0.0050mm, Camber 0.0130mm. NORELL Select Series NMR Tubes are specifically designed for high-resolution NMR. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.