Learn More
Chemglass Life Sciences Tube, NMR, 3mm, Select Series, Without Caps, 80mm Long, High-Throughput, Concentricity +- 0.011mm, Camber +- 0.04mm
SDP
Catalog No. 50872783
Encompass_Preferred
Tube, NMR, 3mm, Select Series, Without Caps, 80mm Long, High-Throughput, Concentricity 0.011mm, Camber 0.04mm. NORELL Select Series NMR Tubes are specifically designed for high-resolution NMR. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.
Catalog No. 50-872-783 Supplier Chemglass Life Sciences Supplier No. CS3HT80
Tube, NMR, 3mm, Select Series, Without Caps, 80mm Long, High-Throughput, Concentricity 0.011mm, Camber 0.04mm. NORELL Select Series NMR Tubes are specifically designed for high-resolution NMR. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.