Learn More
Chemglass Life Sciences Tube, NMR, 5mm, Standard Series, With Caps, 8" Long, 300 MHz, Concentricity +- 0.010mm, Camber +- 0.038mm
SDP
Catalog No. CXR558
Encompass_Preferred
Tube, NMR; Norell; 5mm; Class B Glass; With Caps; Straight; 300 MHz; Length: 8 in.; Clear
Catalog No. CXR558 Supplier Chemglass Life Sciences Supplier No. CXR558
Tube, NMR; Norell; 5mm; Class B Glass; With Caps; Straight; 300 MHz; Length: 8 in.; Clear