Learn More
Chemglass Life Sciences Tube, NMR, 5mm, Standard Series, With Caps, 7" Long, 200 MHz, Concentricity +- 0.010mm, Camber +- 0.040mm
SDP
Catalog No. C505P7
Encompass_Preferred
Tube, NMR, 5mm, Standard Series, With Caps, 7" Long, 200 MHz, Concentricity ¿ 0.010mm, Camber ¿ 0.040mm. NORELL Standard Series NMR Tubes are manufactured out of ASTM Type 1 Class B glass, commonly referred to as N-51A. Applications that are suited for using this type of glass are routine NMR, where samples are run under room temperatures with no thermal gradients, for auto-sampling systems or high throughput NMR. Redesigned from the ground up and with emphasis on quality and precision, Norell has succeeded in virtually eliminating tube wall and bottom variation. It is not recommended to fuse this glass with standard vacuum manifolds and like, since they are generally made of Type 1 Class A glass.
Catalog No. C505P7 Supplier Chemglass Life Sciences Supplier No. C505P7
Tube, NMR, 5mm, Standard Series, With Caps, 7" Long, 200 MHz, Concentricity ¿ 0.010mm, Camber ¿ 0.040mm. NORELL Standard Series NMR Tubes are manufactured out of ASTM Type 1 Class B glass, commonly referred to as N-51A. Applications that are suited for using this type of glass are routine NMR, where samples are run under room temperatures with no thermal gradients, for auto-sampling systems or high throughput NMR. Redesigned from the ground up and with emphasis on quality and precision, Norell has succeeded in virtually eliminating tube wall and bottom variation. It is not recommended to fuse this glass with standard vacuum manifolds and like, since they are generally made of Type 1 Class A glass.