Learn More
Chemglass Life Sciences Tube, NMR, 5mm, Select Series, With Caps, 7" Long, 500 MHz, Amber, Concentricity +- 0.0050mm, Camber +- 0.0130mm
SDP
Catalog No. CS55007A
Encompass_Preferred
Tube, NMR, 5mm, Select Series, With Caps, 7" Long, 500 MHz, Amber, Concentricity 0.0050mm, Camber 0.0130mm. NORELL Select Series Amber NMR Tubes offer photosensitive materials protection from visible and ultraviolet radiation. Typical absorbance (ABS) is between 0.5 and 1.0 from 650 to 300nm and is greater than 1.0 from 300 to 190nm. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.
Catalog No. CS55007A Supplier Chemglass Life Sciences Supplier No. CS55007A
Tube, NMR, 5mm, Select Series, With Caps, 7" Long, 500 MHz, Amber, Concentricity 0.0050mm, Camber 0.0130mm. NORELL Select Series Amber NMR Tubes offer photosensitive materials protection from visible and ultraviolet radiation. Typical absorbance (ABS) is between 0.5 and 1.0 from 650 to 300nm and is greater than 1.0 from 300 to 190nm. These tubes are manufactured out of ASTM Type 1 Class A glass, commonly referred to as Pyrex 7740 (Corning), Duran, or Kimax (Kimble) KG-33 glass. Several key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or any other application where large temperature variations are required in the experiment.