Learn More
Chemglass Life Sciences Tube, NMR, Screw Cap, For 600 MHz, 8" Long
SDP
Catalog No. CS5600SC8
Encompass_Preferred
Tube, NMR, Screw Cap, For 600 MHz, 8" Long. NORELL Screw Cap NMR Tubes have a convenient screw cap access with the security of a PTFE seal. Supplied with open top cap & PTFE septum for quick and clean access with a 22 gauge standard point needle.
Catalog No. CS5600SC8 Supplier Chemglass Life Sciences Supplier No. CS5600SC8
Tube, NMR, Screw Cap, For 600 MHz, 8" Long. NORELL Screw Cap NMR Tubes have a convenient screw cap access with the security of a PTFE seal. Supplied with open top cap & PTFE septum for quick and clean access with a 22 gauge standard point needle.