Learn More
Sklar Instruments Clamp, Tube Occluding Clamp, Sklar 5-1/2 in., Blunt, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501182727 Shop All Sklar Instruments Products
Encompass_Preferred
Clamp, Tube Occluding Clamp, Sklar, 5-1/2 in., Blunt, Straight, Smooth, Stainless Steel, EA. Sklar Presbyterian Tube Occluding Forceps are a ratcheted, finger ring instrument used to control moderate to heavy flow of fluids or blood. They are a heavyweight instrument and can be used with heavy tubing or vessels. The jaws and tips of this instrument are smooth so as not to cause damage to the vessel being occluded. These forceps should not be used on patient tissue. This product is straight with a length of 5-1/2 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-118-2727 Supplier Sklar Instruments Supplier No. 173995
Clamp, Tube Occluding Clamp, Sklar, 5-1/2 in., Blunt, Straight, Smooth, Stainless Steel, EA. Sklar Presbyterian Tube Occluding Forceps are a ratcheted, finger ring instrument used to control moderate to heavy flow of fluids or blood. They are a heavyweight instrument and can be used with heavy tubing or vessels. The jaws and tips of this instrument are smooth so as not to cause damage to the vessel being occluded. These forceps should not be used on patient tissue. This product is straight with a length of 5-1/2 inches. Lifetime warranty against all manufacturer defects.