Learn More
Sklar Instruments Econo Tube Occluding Forceps, Sklar, 7 in., Blunt, Straight, Smooth, Stainless Steel. PK/12.
SDP
Catalog No. 501437669 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Tube Occluding Forceps, Sklar, 7 in., Blunt, Straight, Smooth, Stainless Steel. Econo Tube Occluding Forceps are a ratcheted, finger ring instrument used to control moderate to heavy flow of fluids or blood. They are a heavyweight instrument and can be used with heavy tubing or vessels. The jaws and tips of this instrument are smooth so as not to cause damage if a vessel is being occluded. These forceps should not be used on patient tissue. This product has slighlty wider jaws than Presbyterian occluding forceps. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with a length of 7 inches. Box of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7669 Supplier Sklar Instruments Supplier No. 21598
Econo Tube Occluding Forceps, Sklar, 7 in., Blunt, Straight, Smooth, Stainless Steel. Econo Tube Occluding Forceps are a ratcheted, finger ring instrument used to control moderate to heavy flow of fluids or blood. They are a heavyweight instrument and can be used with heavy tubing or vessels. The jaws and tips of this instrument are smooth so as not to cause damage if a vessel is being occluded. These forceps should not be used on patient tissue. This product has slighlty wider jaws than Presbyterian occluding forceps. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with a length of 7 inches. Box of 12.