Learn More
CELLTREAT Scientific Products TUBE ONLY, 14mL Culture Tube, PP, Non-Sterile, 1000/cs
SDP
Catalog No. NC3433465
Encompass_Preferred
"14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance"
Catalog No. NC3433465 Supplier CELLTREAT Scientific Products Supplier No. 230437
"14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance"