Learn More
CELLTREAT Scientific Products TUBE ONLY, 2.0mL Screw Top Micro Tube, Self-Standing, Grip Band, Sterile, 1000/cs
SDP
Catalog No. NC3433480
Encompass_Preferred
"High temperature resistivity range–shatter/crack resistant and ideal for boilingEasy 3/4 turn to open and closeCaps are available separately in various colors for easy identification and color-codingAll caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and matchGrip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation"
Catalog No. NC3433480 Supplier CELLTREAT Scientific Products Supplier No. 230831S
"High temperature resistivity range–shatter/crack resistant and ideal for boilingEasy 3/4 turn to open and closeCaps are available separately in various colors for easy identification and color-codingAll caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and matchGrip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation"