Learn More
Ace Glass, Inc. Tube only, pressure, 140mL, #25, 38.1mm OD, 20.3cm long, no plug
SDP
Catalog No. 50979809
Encompass_Preferred
140mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Does not include #25 plug. Dimensions: 38.1mm O.D. x 20.3cm long.
Catalog No. 50-979-809 Supplier Ace Glass, Inc. Supplier No. 8648165
140mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Does not include #25 plug. Dimensions: 38.1mm O.D. x 20.3cm long.