Learn More
Ace Glass, Inc. Tube only, pressure, 15mL, #7, 13mm OD, 30.5cm long, no plug
SDP
Catalog No. 50979797
Encompass_Preferred
15mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 13mm O.D. x 30.5cm long.
Catalog No. 50-979-797 Supplier Ace Glass, Inc. Supplier No. 864851
15mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 13mm O.D. x 30.5cm long.