Learn More
Ace Glass, Inc. Tube only, pressure, 210mL, #25, 38.1mm OD, 30.5cm long, no plug
SDP
Supplier:  Ace Glass, Inc. 8648166
Encompass_Preferred
210mL pressure tube with #25 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 38.1mm O.D. x 30.5cm long.
Catalog No. 50-976-209