Learn More
Ace Glass, Inc. Tube only, pressure, 21mL, #7, 19mm OD, 20.3cm long, no plug
SDP
Catalog No. 50974640
Encompass_Preferred
21mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 19mm O.D. x 20.3cm long.
Catalog No. 50-974-640 Supplier Ace Glass, Inc. Supplier No. 864847
21mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 19mm O.D. x 20.3cm long.