Learn More
Ace Glass, Inc. Tube only, pressure, 2mL, #7, 8mm OD, 17.8cm long, no plug
SDP
Catalog No. 50976204
Encompass_Preferred
2mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include #7 plug. Dimensions: 8mm O.D. x 17.8cm long.
Catalog No. 50-976-204 Supplier Ace Glass, Inc. Supplier No. 8648132
2mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include #7 plug. Dimensions: 8mm O.D. x 17.8cm long.