Learn More
Ace Glass, Inc. Tube only, pressure, 300mL, #36, 50mm OD, 30.5cm long, no plug
SDP
Catalog No. 50976218
Encompass_Preferred
300mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Does not include #36 plug. Dimensions: 50mm O.D. x 30.5cm long.
Catalog No. 50-976-218 Supplier Ace Glass, Inc. Supplier No. 8648196
300mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Does not include #36 plug. Dimensions: 50mm O.D. x 30.5cm long.