Learn More
Ace Glass, Inc. Tube only, pressure, 38mL, #15, 25.4mm OD, 20.3cm long, no plug
SDP
Catalog No. 50979794
Encompass_Preferred
38mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 25.4mm O.D. x 20.3cm long.
Catalog No. 50-979-794 Supplier Ace Glass, Inc. Supplier No. 864829
38mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 25.4mm O.D. x 20.3cm long.