Learn More
Ace Glass, Inc. Tube only, pressure, 3mL, #7, 8mm OD, 30.5cm long, no plug
SDP
Catalog No. 50976172
Encompass_Preferred
3mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 8mm O.D. x 30.5cm long.
Catalog No. 50-976-172 Supplier Ace Glass, Inc. Supplier No. 864850
3mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 8mm O.D. x 30.5cm long.