Learn More
Ace Glass, Inc. Tube only, pressure, 90mL, #36, 50mm OD, 10.2cm long, no plug
SDP
Catalog No. NC1590148
Encompass_Preferred
Ace Glass pressure tubes and bottles are the premier glass pressure vessels in the industry. Flat-bottomed bottles are rated at 60psi at room temperature while our tubes are rated at 150psi at as much as 120 degrees C.
Catalog No. NC1590148 Supplier Ace Glass, Inc. Supplier No. 8648190
Ace Glass pressure tubes and bottles are the premier glass pressure vessels in the industry. Flat-bottomed bottles are rated at 60psi at room temperature while our tubes are rated at 150psi at as much as 120 degrees C.