Learn More
Ace Glass, Inc. Tube only, pressure, 9mL, #7, 13mm OD, 20.3cm long, no plug
SDP
Catalog No. 50979796
Encompass_Preferred
9mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 13mm O.D. x 20.3cm long.
Catalog No. 50-979-796 Supplier Ace Glass, Inc. Supplier No. 864846
9mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Does not include plug. Dimensions: 13mm O.D. x 20.3cm long.