Learn More
Chemglass Life Sciences 2mL Craig Tube, Glass Only
SDP
Catalog No. 501532699
Encompass_Preferred
2mL Craig Tube, Glass Only. Used for microscale recrystallizing of small quantities of reaction product. Consists of a glass outer tube with a non-uniform grind to purposely allow leakage.
Catalog No. 50-153-2699 Supplier Chemglass Life Sciences Supplier No. MW7402
2mL Craig Tube, Glass Only. Used for microscale recrystallizing of small quantities of reaction product. Consists of a glass outer tube with a non-uniform grind to purposely allow leakage.