Learn More
CELLTREAT Scientific Products PCR Tube, 0.2mL, Low Retention, Flat Cap, Clear, Non-sterile
SDP
Catalog No. NC3377697
Encompass_Preferred
Ultra-thin tube walls provide uniform and efficient heat transferAvoid sample loss from evaporation with easy open and close lidsAngularly-attached strip caps prevent lids and hinges from interfering with each other when several strips are racked simultaneously230461 and 230462 strips feature easily separable tubes to customize strips230463 strips are designed with robust inter-well linkages, increasing stability and preventing spills230457 tubes feature a low retention surfaceIndividual tubes are available with either frosted flat caps for easier labeling, or optically clear flat caps for RT PCR applicationsCertified RNase/DNase/DNA-free and non-pyrogenic to ensure reliable experimental resultsConveniently packaged in re-sealable bags to minimize risk of contamination
Catalog No. NC3377697 Supplier CELLTREAT Scientific Products Supplier No. 230457
Ultra-thin tube walls provide uniform and efficient heat transferAvoid sample loss from evaporation with easy open and close lidsAngularly-attached strip caps prevent lids and hinges from interfering with each other when several strips are racked simultaneously230461 and 230462 strips feature easily separable tubes to customize strips230463 strips are designed with robust inter-well linkages, increasing stability and preventing spills230457 tubes feature a low retention surfaceIndividual tubes are available with either frosted flat caps for easier labeling, or optically clear flat caps for RT PCR applicationsCertified RNase/DNase/DNA-free and non-pyrogenic to ensure reliable experimental resultsConveniently packaged in re-sealable bags to minimize risk of contamination