Learn More
Thomas Scientific AmplitubeT, Dome Cap, Natural
SDP
Catalog No. 501458974
Encompass_Preferred
Made of polypropylene Simport Reaction Strips include 8 or 12 integral 0.2 ml tubes with ultrathin sidewalls and bottoms for more uniform and efficient temperature transfer, therefore reducing PCR reaction time in most 96-well “V” bottomed thermal cyclers such as MJ Research, Perkin Elmer, Hybaid and others. They are more easily handled than single tubes. They will precisely fit standard well spacing and can also be used with 8- and 12-channel hand-held pipettors. All strips are molded of polypropylene under the most stringent conditions and are offered, colorless and in four different colors. Non-attached cap strips are available in a dome or flat top design and ensure a perfect closure during the whole thermal cycle. Cap strips are not included and have to be ordered separately (see T321 Series). Packed in tamperproof resealable bags. Certified RNase, DNase, Pyrogen and DNA-free
Catalog No. 50-145-8974 Supplier Thomas Scientific Supplier No. 1219F22
Made of polypropylene Simport Reaction Strips include 8 or 12 integral 0.2 ml tubes with ultrathin sidewalls and bottoms for more uniform and efficient temperature transfer, therefore reducing PCR reaction time in most 96-well “V” bottomed thermal cyclers such as MJ Research, Perkin Elmer, Hybaid and others. They are more easily handled than single tubes. They will precisely fit standard well spacing and can also be used with 8- and 12-channel hand-held pipettors. All strips are molded of polypropylene under the most stringent conditions and are offered, colorless and in four different colors. Non-attached cap strips are available in a dome or flat top design and ensure a perfect closure during the whole thermal cycle. Cap strips are not included and have to be ordered separately (see T321 Series). Packed in tamperproof resealable bags. Certified RNase, DNase, Pyrogen and DNA-free