Learn More
Beckman Coulter Beckman Coulter Polyethylene Tube, 400 μL, 7 x 40 mm, Snap-On Cap, 1,000 Pack
Catalog No. NC2380832
Encompass
These polyethylene microcentrifuge tubes are designed for routine laboratory sample handling and benchtop centrifugation. Each tube holds 400 μL and is supplied in bulk 1,000-piece packs for high-volume workflows.
  • Holds 400 μL.
  • Polyethylene construction for chemical compatibility.
  • Dimensions: 7 x 40 mm.
  • Snap-on cap for secure closure.
  • Pack quantity: 1,000 tubes.
  • Suitable for use in benchtop centrifuge rotors.
Catalog No. NC2380832 Supplier Beckman Coulter Supplier No. 314326
These polyethylene microcentrifuge tubes are designed for routine laboratory sample handling and benchtop centrifugation. Each tube holds 400 μL and is supplied in bulk 1,000-piece packs for high-volume workflows.
  • Holds 400 μL.
  • Polyethylene construction for chemical compatibility.
  • Dimensions: 7 x 40 mm.
  • Snap-on cap for secure closure.
  • Pack quantity: 1,000 tubes.
  • Suitable for use in benchtop centrifuge rotors.