Learn More
AB Sciex LLC TUBE PEEKSIL 50UMID 75CML 1/32
Catalog No. NC1109379
Encompass
TUBE PEEKSIL 50UMID 75CML 1/32 ..
Catalog No. NC1109379 Supplier AB Sciex LLC Supplier No. 20500049
TUBE PEEKSIL 50UMID 75CML 1/32 ..