Learn More
Grainger Hydra-Zorb Bronco II Pipe Saddle, Polypropylene Copolymer, 2-1/4 in, 48/Pk.
Catalog No. 502777957
Encompass_Preferred
The Bronco II pipe saddle is designed to securely support tube or pipe runs within strut channel systems. Constructed from durable polypropylene copolymer, it provides a stable mount that minimizes the risk of mold and corrosion. This saddle features an enhanced safety lip that protects insulation against crushing and tearing caused by pipe movement, ensuring long-term integrity of the installation.
  • UL classified 2043 (25/50) for use in plenums and air handling systems
  • Enhanced safety lip prevents insulation damage from pipe movement
  • UV resistant material for extended durability
  • Molded with self-tapping screw location hole
  • Universal mounting feature for secure fit on either side of channel
  • No assembly or tools required for installation
Catalog No. 50-277-7957 Supplier Grainger Supplier No. 193ZL7
The Bronco II pipe saddle is designed to securely support tube or pipe runs within strut channel systems. Constructed from durable polypropylene copolymer, it provides a stable mount that minimizes the risk of mold and corrosion. This saddle features an enhanced safety lip that protects insulation against crushing and tearing caused by pipe movement, ensuring long-term integrity of the installation.
  • UL classified 2043 (25/50) for use in plenums and air handling systems
  • Enhanced safety lip prevents insulation damage from pipe movement
  • UV resistant material for extended durability
  • Molded with self-tapping screw location hole
  • Universal mounting feature for secure fit on either side of channel
  • No assembly or tools required for installation