Learn More
Grainger Hydra-Zorb Tube And Pipe Saddle, Polypropylene Copolymer, White, 4.3 in. Insulation Outside Diameter
Catalog No. 502777952
Encompass_Preferred
The BRONCO II tube and pipe saddle is designed for secure, durable pipe support in insulation systems. Manufactured from high-strength polypropylene copolymer, this white saddle is engineered to resist UV exposure and maintain structural integrity. The design includes an enhanced safety lip to prevent crushing and tearing of insulation caused by pipe movement, ensuring a professional and secure installation.
  • UL 2043 (25/50) classified for use in plenums and air handling systems.
  • Enhanced safety lip prevents insulation damage from pipe movement.
  • Molded with self-tapping screw location hole for easy installation.
  • Extra strength ribbed side walls for added durability.
  • UV resistant material for long-lasting performance.
  • No assembly or tools required for setup.
  • Excellent finished appearance.
  • Manufactured in the U.S.A.
Catalog No. 50-277-7952 Supplier Grainger Supplier No. 193ZM2
The BRONCO II tube and pipe saddle is designed for secure, durable pipe support in insulation systems. Manufactured from high-strength polypropylene copolymer, this white saddle is engineered to resist UV exposure and maintain structural integrity. The design includes an enhanced safety lip to prevent crushing and tearing of insulation caused by pipe movement, ensuring a professional and secure installation.
  • UL 2043 (25/50) classified for use in plenums and air handling systems.
  • Enhanced safety lip prevents insulation damage from pipe movement.
  • Molded with self-tapping screw location hole for easy installation.
  • Extra strength ribbed side walls for added durability.
  • UV resistant material for long-lasting performance.
  • No assembly or tools required for setup.
  • Excellent finished appearance.
  • Manufactured in the U.S.A.