Learn More
Grainger Hydra-Zorb Bronco II Pipe Saddle, Polypropylene Copolymer, 4-1/4 in., Black, 1 ea.
Catalog No. 502777951
Encompass_Preferred
The Bronco® II insulation tube and pipe saddle is designed to provide secure support for insulated pipe systems while preventing damage to the insulation. Manufactured from high-strength polypropylene copolymer, this component features a universal mounting design for easy installation on strut channels. Its construction includes an enhanced safety lip that protects against insulation crushing and tearing during pipe movement.
  • Universal mounting feature allows for a secure fit on either side of the channel
  • Enhanced safety lip prevents insulation crushing and tearing from pipe movement
  • Molded with a self-tapping screw location hole
  • Extra strength ribbed side walls
  • UV resistant and suitable for various environmental conditions
  • No assembly or tools required
  • UL classified 2043 (25/50)
  • Made in the USA
Catalog No. 50-277-7951 Supplier Grainger Supplier No. 193ZM1
The Bronco® II insulation tube and pipe saddle is designed to provide secure support for insulated pipe systems while preventing damage to the insulation. Manufactured from high-strength polypropylene copolymer, this component features a universal mounting design for easy installation on strut channels. Its construction includes an enhanced safety lip that protects against insulation crushing and tearing during pipe movement.
  • Universal mounting feature allows for a secure fit on either side of the channel
  • Enhanced safety lip prevents insulation crushing and tearing from pipe movement
  • Molded with a self-tapping screw location hole
  • Extra strength ribbed side walls
  • UV resistant and suitable for various environmental conditions
  • No assembly or tools required
  • UL classified 2043 (25/50)
  • Made in the USA