Learn More
Grainger Hydra-Zorb Insulation Tube And Pipe Saddle, Polypropylene Copolymer, 5-1/4 in., Black, 12/Pk.
Catalog No. 502778085
Encompass_Preferred
The BRONCO-II insulation tube and pipe saddle is designed to provide secure support for piping systems while protecting insulation. Manufactured from high-strength polypropylene copolymer, this black saddle is engineered for durability and longevity in various environments. It features a design that accommodates pipes and tubing to ensure proper alignment and protection within strut channel systems.
  • Universal mounting feature allows for a secure fit on either side of the channel
  • UL classified 2043 (25/50) for use in plenums and air handling systems
  • Enhanced safety lip prevents insulation crushing and tearing from pipe movement
  • Extra strength ribbed side walls
  • Molded with self-tapping screw location hole
  • No assembly or tools required
  • UV resistant for extended service life
  • Made in the U.S.A.
Catalog No. 50-277-8085 Supplier Grainger Supplier No. 193ZM3
The BRONCO-II insulation tube and pipe saddle is designed to provide secure support for piping systems while protecting insulation. Manufactured from high-strength polypropylene copolymer, this black saddle is engineered for durability and longevity in various environments. It features a design that accommodates pipes and tubing to ensure proper alignment and protection within strut channel systems.
  • Universal mounting feature allows for a secure fit on either side of the channel
  • UL classified 2043 (25/50) for use in plenums and air handling systems
  • Enhanced safety lip prevents insulation crushing and tearing from pipe movement
  • Extra strength ribbed side walls
  • Molded with self-tapping screw location hole
  • No assembly or tools required
  • UV resistant for extended service life
  • Made in the U.S.A.