Learn More
Med Vet International McKesson Test Tube, 5 mL, 13 x 75 mm, 177-110470, 4000/Case
Catalog No. 503044099
Encompass_Preferred
McKesson Test Tube Round Bottom Plain 13 X 75 mm 5 mL Without Color Coding Without Closure Polystyrene Tube 5 mL polystyrene translucent test tubes are rimless 13 x 75 millimeters Molded from premium-grade polystyrene (PS) for clarity Can withstand 3,000 RPM during centrifugation Tubes are nongraduated with round bottoms and no color coding Brittleness temperature: 20° C Maximum temperature: 90° C
Catalog No. 50-304-4099 Supplier Med Vet International Supplier No. 1175263CS
McKesson Test Tube Round Bottom Plain 13 X 75 mm 5 mL Without Color Coding Without Closure Polystyrene Tube 5 mL polystyrene translucent test tubes are rimless 13 x 75 millimeters Molded from premium-grade polystyrene (PS) for clarity Can withstand 3,000 RPM during centrifugation Tubes are nongraduated with round bottoms and no color coding Brittleness temperature: 20° C Maximum temperature: 90° C