Learn More
Ace Glass, Inc. Tube, filter, porosity C (25-50micron), (2)glass stopcock sidearms, 24/40 inner joints, No-Air
SDP
Catalog No. 50979455
Encompass_Preferred
Filter tube for NO-AIR™ applications, with two inner joints. Two, upper and lower side arms with either 2mm stopcocks or a 0-3 inlet valve. Two porosities are standard for the inner fritted disc for stronger and more precise flow.
Catalog No. 50-979-455 Supplier Ace Glass, Inc. Supplier No. 777424
Filter tube for NO-AIR™ applications, with two inner joints. Two, upper and lower side arms with either 2mm stopcocks or a 0-3 inlet valve. Two porosities are standard for the inner fritted disc for stronger and more precise flow.