Learn More
Ace Glass, Inc. Tube, filter, porosity D (10-20micron), (2)inlet valve sidearms, 14/20 inner joints, No-Air
SDP
Catalog No. 50975403
Encompass_Preferred
Filter tube for NO-AIR™ applications, with two inner joints. Two, upper and lower side arms with either 2mm stopcocks or a 0-3 inlet valve. Two porosities are standard for the inner fritted disc for stronger and more precise flow.
Catalog No. 50-975-403 Supplier Ace Glass, Inc. Supplier No. 7774218
Filter tube for NO-AIR™ applications, with two inner joints. Two, upper and lower side arms with either 2mm stopcocks or a 0-3 inlet valve. Two porosities are standard for the inner fritted disc for stronger and more precise flow.