Learn More
MTC Bio FlowTubes™, with standard cap, sterile, 12x75mm, 25/ bag
Catalog No. 501871139
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Made of crystal clear polystyrene, these 12x75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences.The diameter and wall thickness of the tube’s neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. They are available withe the standard two-position culture cap, no cap at all, or with a cap that contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.
Catalog No. 50-187-1139 Supplier MTC Bio Supplier No. T9010
This item has a minimum order quantity of 2 per supplier requirements
Made of crystal clear polystyrene, these 12x75mm FlowTubes™ are designed for Fluorescence Activated Cell Sorting/ flow cytometry equipment including FACS systems from BD Biosciences.The diameter and wall thickness of the tube’s neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. They are available withe the standard two-position culture cap, no cap at all, or with a cap that contains a 35µm nylon mesh screen, designed to assist in cell sorting/straining applications. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.