Learn More
Beckman Coulter Quick-Seal Round Top Polypropylene Tube, 38 x 102mm, 100 mL, 25/pk
Catalog No. NC0695101
Encompass_Preferred
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Quick-Seal tubes provides a secondary BioSafe containment. It also allows you to seal with a simple, hand-held sealing tool without plugs or O-rings. Quick-Seal tube spacer required.
Catalog No. NC0695101 Supplier Beckman Coulter Supplier No. 345776
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Quick-Seal tubes provides a secondary BioSafe containment. It also allows you to seal with a simple, hand-held sealing tool without plugs or O-rings. Quick-Seal tube spacer required.