Learn More
Chemglass Life Sciences Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45°
SDP
Catalog No. 501531642
Encompass_Preferred
Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45. Race tubes are used to measure basal hyphal linear growth as well as circadian conidial banding in the filamentous ascomycete Neurospora crassa.
Catalog No. 50-153-1642 Supplier Chemglass Life Sciences Supplier No. CG402014
Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45. Race tubes are used to measure basal hyphal linear growth as well as circadian conidial banding in the filamentous ascomycete Neurospora crassa.