Learn More
Chemglass Life Sciences Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45°
SDP
Catalog No. 501531643
Encompass_Preferred
Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45. Race tubes are used to measure basal hyphal linear growth as well as circadian conidial banding in the filamentous ascomycete Neurospora crassa.
Catalog No. 50-153-1643 Supplier Chemglass Life Sciences Supplier No. CG402016
Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45. Race tubes are used to measure basal hyphal linear growth as well as circadian conidial banding in the filamentous ascomycete Neurospora crassa.