Learn More
Chemglass Life Sciences Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45°
SDP
Supplier:  Chemglass Life Sciences CG402018
Encompass_Preferred
Race Tube, Glass Only, 16mm OD X 16.5" Overall Length, Both Ends Bent 45. Race tubes are used to measure basal hyphal linear growth as well as circadian conidial banding in the filamentous ascomycete Neurospora crassa.
Catalog No. 50-153-1644
Add to Cart
Request a Quote