Learn More
Research Products International Corp Centrifuge Tube Rack, Capacity 36 x 15ml Tubes, Green, 2 per Package
SDP
Catalog No. 50550216
Encompass
Convenient racks for handling and storing 15ml centrifuge tubes. Constructed of polypropylene that has a working temperature of -80°C to +121°C. Each rack features a permanent indexed grid, two writing surfaces and a clear tight fitting lid. Ideal for organizing freezers or refrigerators.
Catalog No. 50-550-216 Supplier Research Products International Corp Supplier No. 168415
Convenient racks for handling and storing 15ml centrifuge tubes. Constructed of polypropylene that has a working temperature of -80°C to +121°C. Each rack features a permanent indexed grid, two writing surfaces and a clear tight fitting lid. Ideal for organizing freezers or refrigerators.