Learn More
Beckman Coulter Beckman Coulter Tube Adapter, Delrin, for 25 mm thinwall centrifuge tubes (for Type 40.3 rotor)
Catalog No. NC2741858
Encompass
Beckman Coulter Delrin tube adapter (P/N 348123) adapts Type 40.3 rotor cavities to accept thinwall centrifuge tubes with a 25 mm diameter, supporting 33.0 mL and 38.5 mL tube volumes. It provides a secure seating for thinwall polyallomer and ultra-clear tubes during high-speed centrifugation.
  • Delrin construction provides chemical resistance and dimensional stability.
  • Compatible with 25 mm diameter thinwall centrifuge tubes (25 x 83 mm and 25 x 89 mm).
  • Supports 33.0 mL and 38.5 mL tube volumes.
  • Designed for use with Type 40.3 rotor to ensure proper fit and balance.
  • Helps secure thinwall polyallomer and ultra-clear tubes during high-speed runs.
Catalog No. NC2741858 Supplier Beckman Coulter Supplier No. 348123
Beckman Coulter Delrin tube adapter (P/N 348123) adapts Type 40.3 rotor cavities to accept thinwall centrifuge tubes with a 25 mm diameter, supporting 33.0 mL and 38.5 mL tube volumes. It provides a secure seating for thinwall polyallomer and ultra-clear tubes during high-speed centrifugation.
  • Delrin construction provides chemical resistance and dimensional stability.
  • Compatible with 25 mm diameter thinwall centrifuge tubes (25 x 83 mm and 25 x 89 mm).
  • Supports 33.0 mL and 38.5 mL tube volumes.
  • Designed for use with Type 40.3 rotor to ensure proper fit and balance.
  • Helps secure thinwall polyallomer and ultra-clear tubes during high-speed runs.