Learn More
Research Products International Corp Centrifuge Tube Rack, Capacity 16 x 50 ml Tubes, Blue, 2 per Package
SDP
Catalog No. 50550217
Encompass
Convenient racks for handling and storing 50ml centrifuge tubes. Constructed of polypropylene that has a working temperature of -80°C to +121°C. Each rack features a permanent indexed grid, two writing surfaces and a clear tight fitting lid. Ideal for organizing freezers or refrigerators.
Catalog No. 50-550-217 Supplier Research Products International Corp Supplier No. 168450
Convenient racks for handling and storing 50ml centrifuge tubes. Constructed of polypropylene that has a working temperature of -80°C to +121°C. Each rack features a permanent indexed grid, two writing surfaces and a clear tight fitting lid. Ideal for organizing freezers or refrigerators.