Learn More
MTC Bio Tube Rack, fraction collector type, 80 x 1.5mL/2mL, red, pack of 5
Catalog No. NC2017377
Encompass_Preferred
These compact, convenient 80 well racks are designed to keep your tubes neatly and efficiently organized. They are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes. They are made of autoclavable polypropylene and are supplied in a rainbow pack of 5 racks (B, O, R, G, Y) or a single color of your choice.
Catalog No. NC2017377 Supplier MTC Bio Supplier No. R1040R
These compact, convenient 80 well racks are designed to keep your tubes neatly and efficiently organized. They are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes. They are made of autoclavable polypropylene and are supplied in a rainbow pack of 5 racks (B, O, R, G, Y) or a single color of your choice.